4-Chloro-3-nitrobenzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12648-22
A12648-06
A12648-14
CAAAAA12648-22EA
723.64
CAD
CAAAAA12648-22
CAAAA12648-06
CAAAA12648-14
4-Chloro-3-nitrobenzonitrile 98%
4-Chloro-3-nitrobenzonitrile
Purity: 98%
CAS No.: 939-80-0
Molecular Formula: C7H3ClN2O2
Formula Weight: 182.57
Melting Point: 98-102°
Beilstein Reference: 1639111
Formula:
C₇H₃ClN₂O₂ MW: 182.57 g/mol Melting Pt: 98…102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016987 CAS Number: 939-80-0 EINECS: 213-364-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...