Triethylamine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(Diethylamino)ethane, TEA
A12646-AK
A12646-0F
A12646-AP
CAAAAA12646-AKEA
60.99
CAD
CAAAAA12646-AK
CAAAA12646-0F
CAAAA12646-AP
Triethylamine 99%
Triethylamine
Purity: 99%
CAS No.: 121-44-8
Molecular Formula: C6H15N
Formula Weight: 101.19
Flash Point: -11°(12°F)
Refractive Index: 1.4005
Odor: Ammonia-like
Melting Point: -115°
Boiling Point: 89-90°
Beilstein Reference: 1843166
Merck Reference: 14,9666
UN No.: 1296
MDL No.: MFCD00009051
Formula:
(C₂H₅)₃N MW: 101.19 g/mol Boiling Pt: 90 Melting Pt: -115 Density: 0.726 Flash Pt: –11 °C (12 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009051 CAS Number: 121-44-8 EINECS: 204-469-4 UN: 1296 ADR: 3 II Merck Index: 14,9666 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...