Trimethylsulfonium iodide 99%

Supplier: Thermo Scientific Chemicals
Warning

A12639-0B A12639-18 A12639-30
CAAAA12639-0BEA 1401.46 CAD
CAAAA12639-0B CAAAA12639-18 CAAAA12639-30
Trimethylsulfonium iodide 99%
Trimethylsulphonium iodide

Purity: 99% CAS No.: 2181-42-2 Molecular Formula: (CH3)3SI Formula Weight: 204.07 Melting Point: ca 215° subl. Beilstein Reference: 3555192

Formula: C₃H₉IS
Melting Pt: ∼ 215 °C (subl.)
Density: 1.922 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011632
CAS Number: 2181-42-2
EINECS: 218-555-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR