1-Bromo-4-chloro-2-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

A12635-22 A12635-06 A12635-14
CAAAAA12635-22EA 477.16 CAD
CAAAAA12635-22 CAAAA12635-06 CAAAA12635-14
1-Bromo-4-chloro-2-nitrobenzene 98%
1-Bromo-4-chloro-2-nitrobenzene

Purity: 98% CAS No.: 41513-04-6 Molecular Formula: C6H3BrClNO2 Formula Weight: 236.45 Melting Point: 68-72° Beilstein Reference: 1950424

Formula: C₆H₃BrClNO₂
Melting Pt: 68…72 °C
Density: 2.048 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00024320
CAS Number: 41513-04-6
EINECS: 255-421-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR