Indole-3-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A12634-03 A12634-06 A12634-14
CAAAA12634-03EA 44.56 CAD
CAAAA12634-03 CAAAA12634-06 CAAAA12634-14
Indole-3-carboxylic acid 98%
Indole-3-carboxylic acid

Purity: 98% CAS No.: 771-50-6 Molecular Formula: C9H7NO2 Formula Weight: 161.16 Melting Point: ca 239° dec. Beilstein Reference: 129435

Formula: C₉H₇NO₂
Melting Pt: ∼ 239 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005624
CAS Number: 771-50-6
EINECS: 212-231-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR