2,3-Dimethylpyrazine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12628-09
A12628-18
CAAAA12628-09EA
64.62
CAD
CAAAA12628-09
CAAAA12628-18
2,3-Dimethylpyrazine 99%
2,3-Dimethylpyrazine
Purity: 99%
CAS No.: 5910-89-4
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 54°(129°F)
Refractive Index: 1.5070
Odor: Pleasant
Melting Point: -12°
Boiling Point: 156°
Beilstein Reference: 107908
UN No.: 1993
MDL No.: MFCD00006144
Formula:
C₆H₈N₂ Boiling Pt: 156 °C Melting Pt: –12 °C Density: 1.022 Flash Pt: 54 °C (129 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006144 CAS Number: 5910-89-4 EINECS: 227-630-0 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...