Butyrylthiocholine iodide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
S-n-Butyrylthiocholine iodide, (2-Butyrylthioethyl)trimethylammonium iodide, 2-(Butyrylthio)-N,N,N-trimethylethan-1-aminium iodide, (Propylcarbonylthioethyl)trimethylammonium iodide, (2-(Butyrylthio)ethyl)trimethylammonium iodide
A12622-06
A12622-14
CAAAA12622-06EA
79.63
CAD
CAAAA12622-06
CAAAA12622-14
Butyrylthiocholine iodide 98%
Butyrylthiocholine iodide
Purity: 98%
CAS No.: 1866-16-6
Molecular Formula: C9H20INOS
Formula Weight: 317.23
Storage Temperature: 2°C to 8°C
Melting Point: 172-176°
Beilstein Reference: 3729509
MDL No.: MFCD00011845
Formula:
H₃C(CH₂)₂C(S)O(CH₂)₂N(CH₃)₃I MW: 317.23 g/mol Melting Pt: 172 Storage Temperature: Refrigerator |
MDL Number:
MFCD00011845 CAS Number: 1866-16-6 EINECS: 217-475-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...