(±)-Camphor-10-sulfonic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12620-22
A12620-36
CAAAA12620-22EA
76
CAD
CAAAA12620-22
CAAAA12620-36
(±)-Camphor-10-sulfonic acid 98%
(±)-Camphor-10-sulphonic acid
Purity: 98%
CAS No.: 5872-08-2
Molecular Formula: C10H16O4S
Formula Weight: 232.30
Melting Point: ca 200° dec.
Beilstein Reference: 3205973
UN No.: 3261
MDL No.: MFCD00074827
Formula:
C₁₀H₁₆O₄S Melting Pt: ca 200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00074827 CAS Number: 5872-08-2 EINECS: 227-527-0 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...