Pyrrole 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1H-Pyrrole
A12616-0B
A12616-18
A12616-30
CAAAA12616-0BEA
883.22
CAD
CAAAA12616-0B
CAAAA12616-18
CAAAA12616-30
Pyrrole 98+%
Pyrrole
Purity: 98+%
CAS No.: 109-97-7
Molecular Formula: C4H5N
Formula Weight: 67.09
Storage Temperature: 2°C to 8°C
Flash Point: 38°(100°F)
Refractive Index: 1.5090
Odor: Mild
Boiling Point: 131°
Beilstein Reference: 1159
Merck Reference: 14,8014
UN No.: 1992
MDL No.: MFCD00005216
Formula:
C₄H₅N MW: 67.09 g/mol Boiling Pt: 131 °C Melting Pt: –24 °C Density: 0.967 Flash Pt: 38 °C (100 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005216 CAS Number: 109-97-7 EINECS: 203-724-7 UN: 1992 ADR: 3,III Merck Index: 14,8014 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...