2-Fluoroaniline 99%

Supplier: Thermo Scientific Chemicals
Warning

A12612-14 A12612-22 A12612-36
CAAAA12612-14EA 42.82 CAD
CAAAA12612-14 CAAAA12612-22 CAAAA12612-36
2-Fluoroaniline 99%
2-Fluoroaniline

Purity: 99% CAS No.: 348-54-9 Molecular Formula: C6H6FN Formula Weight: 111.12 Flash Point: 60°(140°F) Refractive Index: 1.5420 Odor: Recognizable Melting Point: -29° Boiling Point: 171-173° Beilstein Reference: 1524219 UN No.: 2941 MDL No.: MFCD00007642

Formula: C₆H₆FN
Boiling Pt: 171…173 °C
Melting Pt: –29 °C
Density: 1.152
Flash Pt: 60 °C (140 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007642
CAS Number: 348-54-9
EINECS: 206-478-9
UN: 2941
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR