3-Bromobenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12606-14
A12606-22
CAAAA12606-14EA
97.01
CAD
CAAAA12606-14
CAAAA12606-22
3-Bromobenzoic acid 98+%
3-Bromobenzoic acid
Purity: 98+%
CAS No.: 585-76-2
Molecular Formula: C7H5BrO2
Formula Weight: 201.02
Melting Point: 153-157°
Beilstein Reference: 2041792
Formula:
C₇H₅BrO₂ Boiling Pt: 308 °C (1013 hPa) Melting Pt: 153…157 °C Density: 1.948 Storage Temperature: Ambient |
MDL Number:
MFCD00002487 CAS Number: 585-76-2 EINECS: 209-562-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...