1-Methylindole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12605-09
A12605-18
CAAAA12605-09EA
69.05
CAD
CAAAA12605-09
CAAAA12605-18
1-Methylindole 98%
1-Methylindole
Purity: 98%
CAS No.: 603-76-9
Molecular Formula: C9H9N
Formula Weight: 131.18
Flash Point:
Formula:
C₉H₉N Boiling Pt: 237…239 °C Melting Pt: 94…95 °C Density: 1.048 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005800 CAS Number: 603-76-9 EINECS: 210-057-5 UN: 3334 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...