3,4-Difluorophenol 98%

Supplier: Thermo Scientific Chemicals
Danger

A12596-06 A12596-14 A12596-22
CAAAA12596-06EA 86.27 CAD
CAAAA12596-06 CAAAA12596-14 CAAAA12596-22
3,4-Difluorophenol 98%
3,4-Difluorophenol

Purity: 98% CAS No.: 2713-33-9 Molecular Formula: C6H4F2O Formula Weight: 130.09 Flash Point: 57°(134°F) Odor: Phenol-like Boiling Point: 84-86°/20mm Beilstein Reference: 2081085 UN No.: 2921 MDL No.: MFCD00010315

Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 84…86 °C/20 mm
Melting Pt: 34…38 °C
Flash Pt: 57 °C (134 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010315
CAS Number: 2713-33-9
EINECS: 424-300-1
UN: 2925
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR