Acetamide 99%

Supplier: Thermo Scientific Chemicals
Warning

A12589-0B A12589-0I A12589-30
CAAAA12589-0BEA 191.18 CAD
CAAAA12589-0B CAAAA12589-0I CAAAA12589-30
Acetamide 99%
Acetamide

Purity: 99% CAS No.: 60-35-5 Molecular Formula: CH3CONH2 Formula Weight: 59.07 Melting Point: 76-81° Boiling Point: 220-222° Merck Reference: 14,43 MDL No.: MFCD00008023

Formula: C₂H₅NO
Boiling Pt: 220…222 °C
Melting Pt: 76…81 °C
Density: 1.159
Storage Temperature: Ambient
MDL Number: MFCD00008023
CAS Number: 60-35-5
EINECS: 200-473-5
Merck Index: 14,43

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR