Diphenyl disulfide 98%

Supplier: Thermo Scientific Chemicals
Warning

A12586-18 A12586-30
CAAAA12586-18EA 86.58 CAD
CAAAA12586-18 CAAAA12586-30
Diphenyl disulfide 98%
Diphenyl disulphide

Purity: 98% CAS No.: 882-33-7 Molecular Formula: C12H10S2 Formula Weight: 218.34 Odor: Unpleasant Melting Point: 58-62° Boiling Point: 310° Beilstein Reference: 639794 MDL No.: MFCD00003065

Formula: C₁₂H₁₀S₂
Boiling Pt: 310 °C
Melting Pt: 58…62 °C
Density: 1.155 g/cm³ (64.9 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003065
CAS Number: 882-33-7
EINECS: 212-926-4
UN: 3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR