Benzil 98%

Supplier: Thermo Scientific Chemicals
Warning

A12579-0E A12579-22 A12579-36
CAAAA12579-0EEA 518.24 CAD
CAAAA12579-0E CAAAA12579-22 CAAAA12579-36
Benzil 98%
Benzil

Purity: 98% CAS No.: 134-81-6 Molecular Formula: C14H10O2 Formula Weight: 210.23 Melting Point: 94-96° Boiling Point: 346-348° Merck Reference: 14,1078 MDL No.: MFCD00003080

Formula: (C₆H₅)C(O)C(O)(C₆H₅)
Boiling Pt: 346…348 °C
Melting Pt: 94…96 °C
Density: 1.521
Storage Temperature: Ambient
MDL Number: MFCD00003080
CAS Number: 134-81-6
EINECS: 205-157-0
Merck Index: 14,1078

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR