4-Fluorophenyl isocyanate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12576-06
A12576-14
CAAAA12576-06EA
64.31
CAD
CAAAA12576-06
CAAAA12576-14
4-Fluorophenyl isocyanate 98+%
4-Fluorophenyl isocyanate
Purity: 98+%
CAS No.: 1195-45-5
Molecular Formula: C7H4FNO
Formula Weight: 137.11
Storage Temperature: 2°C to 8°C
Flash Point: 52°(125°F)
Refractive Index: 1.5140
Odor: Pungent, makes eyes water
Boiling Point: 55-56°/8mm
Beilstein Reference: 471609
UN No.: 3080
MDL No.: MFCD00002023
Formula:
C₇H₄FNO Boiling Pt: 55…56 °C/8 mm Density: 1.210 Flash Pt: 52 °C (125 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002023 CAS Number: 1195-45-5 EINECS: 214-799-0 UN: 3080 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...