1-Methylimidazole (NMI) 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12575-A3
A12575-22
A12575-36
CAAAAA12575-A3EA
592.5
CAD
CAAAAA12575-A3
CAAAA12575-22
CAAAA12575-36
1-Methylimidazole (NMI) 99%
1-Methylimidazole
Purity: 99%
CAS No.: 616-47-7
Molecular Formula: C4H6N2
Formula Weight: 82.11
Flash Point: 92°(197°F)
Refractive Index: 1.4970
Melting Point: -6°
Boiling Point: 195-197°
Beilstein Reference: 105197
UN No.: 3267
MDL No.: MFCD00005292
Formula:
C₄H₆N₂ Boiling Pt: 195…197 °C Melting Pt: –6 °C Density: 1.031 Flash Pt: 92 °C (197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005292 CAS Number: 616-47-7 EINECS: 210-484-7 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...