2-Methyl-4(5)-nitroimidazole 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Methyl-4-nitro-1H-imidazole, 2-Methyl-5-nitroimidazole, 2-Methyl-5-nitro-1H-imidazole, 2-Methyl-4-nitroimidazole
A12569-18
A12569-30
CAAAA12569-0BEA
590.11
CAD
CAAAA12569-0B
CAAAA12569-18
CAAAA12569-30
2-Methyl-4(5)-nitroimidazole 99%
2-Methyl-4(5)-nitroimidazole
Purity: 99%
CAS No.: 696-23-1
Molecular Formula: C4H5N3O2
Formula Weight: 127.10
Melting Point: 251-254°
Beilstein Reference: 4032
Formula:
C₄H₅N₃O MW: 127.10 g/mol Melting Pt: 251…254 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005191 CAS Number: 696-23-1 EINECS: 211-790-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...