4-Hexylresorcinol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hexylresorcinol, 4-Hexylbenzene-1,3-diol

A12552-06 A12552-14 A12552-22
CAAAA12552-06EA 36.97 CAD
CAAAA12552-06 CAAAA12552-14 CAAAA12552-22
4-Hexylresorcinol 99%
4-Hexylresorcinol

Purity: 99% CAS No.: 136-77-6 Molecular Formula: C12H18O2 Formula Weight: 194.27 Flash Point: 100°(230°F) Melting Point: 64-67° Boiling Point: 333-335° Beilstein Reference: 2048312 Merck Reference: 14,4712 MDL No.: MFCD00002284

Formula: C₁₂H₁₈O₂
Boiling Pt: 333…335 °C
Melting Pt: 64…67 °C
Flash Pt: >100 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002284
CAS Number: 136-77-6
EINECS: 205-257-4
Merck Index: 14,4712

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR