2'-Hydroxyacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12550-22
A12550-36
CAAAA12550-0EEA
589.94
CAD
CAAAA12550-0E
CAAAA12550-22
CAAAA12550-36
2'-Hydroxyacetophenone 98%
2'-Hydroxyacetophenone
Purity: 98%
CAS No.: 118-93-4
Molecular Formula: C8H8O2
Formula Weight: 136.15
Flash Point: 106°(222°F)
Refractive Index: 1.5584
Melting Point: 3-6°
Boiling Point: 218°
Beilstein Reference: 386123
MDL No.: MFCD00002219
Formula:
C₈H₈O₂ Boiling Pt: 218 °C Melting Pt: 3…6 °C Density: 1.130 Flash Pt: 106 °C(222 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002219 CAS Number: 118-93-4 EINECS: 204-288-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...