2,4-Dimethoxybenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12549-14
A12549-22
CAAAA12549-14EA
64.31
CAD
CAAAA12549-14
CAAAA12549-22
CAAAA12549-36
2,4-Dimethoxybenzaldehyde 98%
2,4-Dimethoxybenzaldehyde
Purity: 98%
CAS No.: 613-45-6
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 67-71°
Boiling Point: 165°/10mm
MDL No.: MFCD00003311
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Boiling Pt: 165 °C/10 mm Melting Pt: 67…71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003311 CAS Number: 613-45-6 EINECS: 210-342-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...