N,N-Dimethyloctylamine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12546-30
A12546-09
A12546-18
CAAAAA12546-30EA
811.42
CAD
CAAAAA12546-30
CAAAA12546-09
CAAAA12546-18
N,N-Dimethyloctylamine 95%
N,N-Dimethyloctylamine
Purity: 95%
CAS No.: 7378-99-6
Molecular Formula: CH3(CH2)7N(CH3)2
Formula Weight: 157.30
Flash Point: 65°(149°F)
Refractive Index: 1.4243
Melting Point: -57°
Boiling Point: 195°
Beilstein Reference: 1698081
UN No.: 2922
MDL No.: MFCD00009558
Formula:
C₁₀H₂₃N Boiling Pt: 195 °C Melting Pt: –57 °C Density: 0.765 Flash Pt: 65 °C (149 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009558 CAS Number: 7378-99-6 EINECS: 230-939-3 UN: 2922 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...