Ethyl acetoacetate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12544-30
A12544-0C
A12544-0E
A12544-36
CAAAAA12544-30EA
45.35
CAD
CAAAAA12544-30
CAAAA12544-0C
CAAAA12544-0E
CAAAA12544-36
Ethyl acetoacetate 99+%
Ethyl acetoacetate
Purity: 99+%
CAS No.: 141-97-9
Molecular Formula: C6H10O3
Formula Weight: 130.14
Flash Point: 84°(183°F)
Physical Form: Liquid
Density: 1.025
Refractive Index: 1.4190
Appearance: Colorless
Odor: Fruit-like
Melting Point: -43°
Boiling Point: 180-181°
Beilstein Reference: 385838
Merck Reference: 14,3758
MDL No.: MFCD00009199
Formula:
C₆H₁₀O₃ Boiling Pt: 180…181 °C Melting Pt: –43 °C Density: 1.025 Flash Pt: 84 °C(183 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009199 CAS Number: 141-97-9 EINECS: 205-516-1 Merck Index: 14,3758 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...