4-Nitrobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-Nitrobenzoyl chloride

A12543-09 A12543-0B A12543-30
CAAAAA12543-09EA 41.24 CAD
CAAAAA12543-09 CAAAA12543-0B CAAAA12543-30
4-Nitrobenzoyl chloride 98%
4-Nitrobenzoyl chloride

Purity: 98% CAS No.: 122-04-3 Molecular Formula: C7H4ClNO3 Formula Weight: 185.57 Flash Point: 102°(215°F) Odor: Characteristic Boiling Point: 155°/15mm Beilstein Reference: 473192 Merck Reference: 14,6590 UN No.: 3261 MDL No.: MFCD00007345

Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Boiling Pt: 278
Melting Pt: 71…74 °C
Density: 1.53
Flash Pt: 102 °C (215 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007345
CAS Number: 122-04-3
EINECS: 204-517-4
UN: 3261
ADR: 8 II
Merck Index: 14,6590

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR