3'-Chloropropiophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

A12542-14 A12542-22 A12542-36
CAAAA12542-14EA 59.25 CAD
CAAAA12542-14 CAAAA12542-22 CAAAA12542-36
3'-Chloropropiophenone 98%
3'-Chloropropiophenone

Purity: 98% CAS No.: 34841-35-5 Molecular Formula: C9H9ClO Formula Weight: 168.62 Flash Point: 110°(230°F) Melting Point: 45-47° Boiling Point: 123-124°/13mm MDL No.: MFCD00009925

Formula: C₉H₉ClO
Boiling Pt: 123…124 °C/13 mm
Melting Pt: 45…47 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009925
CAS Number: 34841-35-5
EINECS: 252-242-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR