Phenoxyacetic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12541-22
A12541-0E
A12541-36
CAAAAA12541-22EA
39.97
CAD
CAAAAA12541-22
CAAAA12541-0C
CAAAA12541-0E
CAAAA12541-36
Phenoxyacetic acid 99%
Phenoxyacetic acid
Purity: 99%
CAS No.: 122-59-8
Molecular Formula: C8H8O3
Formula Weight: 152.15
Flash Point: 165°(329°F)
Melting Point: 98-101°
Boiling Point: 285°
Beilstein Reference: 907949
Merck Reference: 14,7254
MDL No.: MFCD00004296
Formula:
C₈H₈O₃ Boiling Pt: 285 °C Melting Pt: 98…101 °C Density: 1.36 g/cm³ (20 °C) Flash Pt: 165 °C(329 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004296 CAS Number: 122-59-8 EINECS: 204-556-7 Merck Index: 14,7254 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...