Ethyl-4-chlorobenzoate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12540-18
A12540-30
CAAAA12540-0BEA
1052.96
CAD
CAAAA12540-0B
CAAAA12540-18
CAAAA12540-30
Ethyl-4-chlorobenzoate 98+%
Ethyl 4-chlorobenzoate
Purity: 98+%
CAS No.: 7335-27-5
Molecular Formula: C9H9ClO2
Formula Weight: 184.62
Refractive Index: 1.5240
Boiling Point: 237-239°
MDL No.: MFCD00013645
Formula:
C₉H₉ClO₂ Boiling Pt: 237…239 °C Density: 1.18 Storage Temperature: Ambient |
MDL Number:
MFCD00013645 CAS Number: 7335-27-5 EINECS: 230-844-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...