Trimethylsilyl trifluoromethanesulfonate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trimethylsilyl trifluoromethanesulfonate, Trimethylsilyl trifluoromethanesulphonate
A12535-09
A12535-18
A12535-30
CAAAA12535-09EA
73.15
CAD
CAAAA12535-09
CAAAA12535-18
CAAAA12535-30
Trimethylsilyl trifluoromethanesulfonate 99%
Trimethylsilyl trifluoromethanesulphonate
Purity: 99%
CAS No.: 27607-77-8
Molecular Formula: (CH3)3SiSO3CF3
Formula Weight: 222.26
Flash Point: 25°(77°F)
Refractive Index: 1.3600
Boiling Point: 39-40°/12mm
Beilstein Reference: 1868911
Merck Reference: 14,9719
UN No.: 2924
MDL No.: MFCD00000406
Formula:
C₄H₉F₃O₃SSi Boiling Pt: 39…40 °C/12 mm Density: 1.225 Flash Pt: 25 °C (77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000406 CAS Number: 27607-77-8 EINECS: 248-565-4 UN: 2920 ADR: 8,II Merck Index: 14,9719 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...