(1-Pentyl)triphenylphosphonium bromide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pentyltriphenylphosphonium bromide

A12534-14 A12534-22 A12534-36
CAAAA12534-14EA 71.26 CAD
CAAAA12534-14 CAAAA12534-22 CAAAA12534-36
(1-Pentyl)triphenylphosphonium bromide 98%
(1-Pentyl)triphenylphosphonium bromide

Purity: 98% CAS No.: 21406-61-1 Molecular Formula: CH3(CH2)4P(C6H5)3Br Formula Weight: 413.33 Melting Point: 164-166° Beilstein Reference: 3580453

Formula: H₃C(CH₂)₄P(C₆H₅)₃Br
MW: 413.34 g/mol
Melting Pt: 168
Storage Temperature: Ambient
MDL Number: MFCD00031630
CAS Number: 21406-61-1
EINECS: 244-374-5
UN: 3464
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2bf324e98e27624d1eab0e430f6eee7a96fa960&vid=e2bf324e98e27624d1eab0e430f6eee7a96fa960&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR