(4-Chlorophenyl)acetyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

A12530-30 A12530-09 A12530-18
CAAAAA12530-30EA 1215.02 CAD
CAAAAA12530-30 CAAAA12530-09 CAAAA12530-18
(4-Chlorophenyl)acetyl chloride 98%
(4-Chlorophenyl)acetyl chloride

Purity: 98% CAS No.: 25026-34-0 Molecular Formula: C8H6Cl2O Formula Weight: 189.04 Flash Point: 110°(230°F) Refractive Index: 1.5510 Boiling Point: 112-114°/11mm Beilstein Reference: 637775 UN No.: 3265 MDL No.: MFCD00037111

Formula: (C₆H₄Cl)CH₂COCl
Boiling Pt: 112…114 °C/11 mm
Density: 1.292
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00037111
CAS Number: 25026-34-0
EINECS: 246-571-1
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR