Ethyl crotonate 98%

Supplier: Thermo Scientific Chemicals
Warning

A12529-0E A12529-22 A12529-36
CAAAA12529-0EEA 774.2 CAD
CAAAA12529-0E CAAAA12529-22 CAAAA12529-36
Ethyl crotonate 98%
Ethyl crotonate

Purity: 98% CAS No.: 623-70-1 Molecular Formula: CH3CH=CHCO2CH2CH3 Formula Weight: 114.14 Flash Point: 2°(36°F) Boiling Point: 141-143° UN No.: 1862 MDL No.: MFCD00009289

Formula: C₆H₁₀O₂
Boiling Pt: 141…143 °C
Melting Pt: –79 °C
Density: 0.918
Flash Pt: 2 °C (36 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009289
CAS Number: 623-70-1
EINECS: 210-808-7
UN: 1862
ADR: 3,II
Merck Index: 13,02625

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR