Phenethyl bromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2-Bromoethyl)benzene, 2-Phenylethyl bromide
A12528-22
A12528-36
CAAAA12528-22EA
57.83
CAD
CAAAA12528-22
CAAAA12528-36
Phenethyl bromide 98%
Phenethyl bromide
Purity: 98%
CAS No.: 103-63-9
Molecular Formula: C8H9Br
Formula Weight: 185.06
Flash Point: 89°(192°F)
Refractive Index: 1.5560
Melting Point: -56°
Boiling Point: 220-221°
Beilstein Reference: 507487
MDL No.: MFCD00000240
Formula:
BrCH₂CH₂C₆H₅ MW: 185.06 g/mol Boiling Pt: 221 Melting Pt: -56 Density: 1.355 Flash Pt: 89 °C (192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000240 CAS Number: 103-63-9 EINECS: 203-130-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...