Terephthalic acid 98+%

Supplier: Thermo Scientific Chemicals

A12527-30 A12527-36
CAAAAA12527-30EA 47.02 CAD
CAAAAA12527-30 CAAAA12527-0C CAAAA12527-36
Terephthalic acid 98+%
Terephthalic acid

Purity: 98+% CAS No.: 100-21-0 Molecular Formula: C8H6O4 Formula Weight: 166.13 Flash Point: 260°(500°F) Odor: Odorless Beilstein Reference: 1909333 Merck Reference: 14,9162 MDL No.: MFCD00002558

Formula: C₈H₆O₄
Boiling Pt: 392 °C (1013 hPa)
Melting Pt: 402 °C (subl.)
Density: 1.510
Flash Pt: 260 °C(500 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002558
CAS Number: 100-21-0
EINECS: 202-830-0
Merck Index: 14,9162

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR