2,4-Dichlorobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12525-0E
A12525-22
A12525-36
CAAAA12525-0EEA
902.18
CAD
CAAAA12525-0E
CAAAA12525-22
CAAAA12525-36
2,4-Dichlorobenzaldehyde 98%
2,4-Dichlorobenzaldehyde
Purity: 98%
CAS No.: 874-42-0
Molecular Formula: C7H4Cl2O
Formula Weight: 175.01
Flash Point: 127°(260°F)
Melting Point: 69-73°
Boiling Point: 233°
Beilstein Reference: 471601
UN No.: 1759
MDL No.: MFCD00003305
Formula:
C₇H₄Cl₂O Boiling Pt: 233 °C Melting Pt: 69…73 °C Density: 1.33 g/cm³ (20 °C) Flash Pt: 127 °C(260 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003305 CAS Number: 874-42-0 EINECS: 212-861-1 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...