4-Bromobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A12518-36 A12518-14 A12518-22
CAAAAA12518-36EA 935.36 CAD
CAAAAA12518-36 CAAAA12518-14 CAAAA12518-22
4-Bromobenzoic acid 98+%
4-Bromobenzoic acid

Purity: 98+% CAS No.: 586-76-5 Molecular Formula: C7H5BrO2 Formula Weight: 201.02 Odor: Odorless Melting Point: 253-258° Beilstein Reference: 1906923 Merck Reference: 14,1408 MDL No.: MFCD00002529

Formula: C₇H₅BrO₂
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 253…258 °C
Density: 1.894
Storage Temperature: Ambient
MDL Number: MFCD00002529
CAS Number: 586-76-5
EINECS: 209-581-7
Merck Index: 14,1408

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR