Phenothiazine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12517-36
A12517-0I
A12517-30
CAAAAA12517-36EA
80.11
CAD
CAAAAA12517-36
CAAAA12517-0I
CAAAA12517-30
Phenothiazine 98+%
Phenothiazine
Acts on type I calcium/calmodulin dependent phosphodiesterase
Purity: 98+%
CAS No.: 92-84-2
Molecular Formula: C12H9NS
Formula Weight: 199.28
Melting Point: 182-186°
Boiling Point: 235°/21mm
Merck Reference: 14,7252
MDL No.: MFCD00005015
Formula:
C₁₂H₉NS Boiling Pt: 235 °C/21 mm Melting Pt: 182…186 °C Density: 1.34 g/cm³ (20 °C) Flash Pt: 202 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005015 CAS Number: 92-84-2 EINECS: 202-196-5 UN: 3077 ADR: 9,III Merck Index: 14,7252 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...