2-Thenoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A12514-14 A12514-22 A12514-36
CAAAA12514-14EA 40.45 CAD
CAAAA12514-14 CAAAA12514-22 CAAAA12514-36
2-Thenoic acid 99%
2-Thenoic acid

A hypoglycemic, anti-lipolytic agent


Purity: 99% CAS No.: 527-72-0 Molecular Formula: C5H4O2S Formula Weight: 128.15 Melting Point: 126-130° Boiling Point: 259-261° Merck Reference: 14,9354 MDL No.: MFCD00005437

Formula: C₅H₄O₂S
Boiling Pt: 259…261 °C
Melting Pt: 126…130 °C
Density: 1.42
Storage Temperature: Ambient
MDL Number: MFCD00005437
CAS Number: 527-72-0
EINECS: 208-423-4
Merck Index: 14,9354

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR