1-(Trimethylsilyl)imidazole 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12512-06
A12512-22
A12512-18
A12512-30
CAAAAA12512-06EA
39.5
CAD
CAAAAA12512-06
CAAAAA12512-22
CAAAA12512-18
CAAAA12512-30
1-(Trimethylsilyl)imidazole 97%
1-(Trimethylsilyl)imidazole
Purity: 97%
CAS No.: 18156-74-6
Molecular Formula: C6H12N2Si
Formula Weight: 140.26
Flash Point: 18°(64°F)
Refractive Index: 1.4765
Odor: Weak
Melting Point: -42°
Boiling Point: 212°
Beilstein Reference: 606148
UN No.: 1993
MDL No.: MFCD00005280
Formula:
C₆H₁₂N₂Si Boiling Pt: 212 °C Melting Pt: –42 °C Density: 0.956 Flash Pt: 18 °C (64 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005280 CAS Number: 18156-74-6 EINECS: 242-040-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...