1-Nonanol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Nonan-1-ol, n-Nonanol, 1-Nonyl alcohol, Nonyl alcohol, 1-Hydroxy nonane, n-Nonyl alcohol, Octyl carbinol

A12510-22 A12510-36
CAAAA12510-22EA 70.94 CAD
CAAAA12510-22 CAAAA12510-36
1-Nonanol 99%
1-Nonanol

Purity: 99% CAS No.: 143-08-8 Molecular Formula: C9H20O Formula Weight: 144.26 Flash Point: 75°(167°F) Refractive Index: 1.4330 Odor: Pleasant Melting Point: -7° Boiling Point: 214-216° Beilstein Reference: 969213 Merck Reference: 14,6679 UN No.: 3082 MDL No.: MFCD00002990

Formula: H₃C(CH₂)₇CH₂OH
MW: 144.26 g/mol
Boiling Pt: 215
Melting Pt: -7
Density: 0.827
Flash Pt: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002990
CAS Number: 143-08-8
EINECS: 205-583-7
UN: 3082
ADR: 9 III
Merck Index: 14,6679

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR