3-Fluorobenzyl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12509-14
A12509-09
A12509-18
CAAAAA12509-14EA
243.32
CAD
CAAAAA12509-14
CAAAA12509-09
CAAAA12509-18
3-Fluorobenzyl alcohol 98%
3-Fluorobenzyl alcohol
Purity: 98%
CAS No.: 456-47-3
Molecular Formula: C7H7FO
Formula Weight: 126.13
Flash Point: 90°(194°F)
Refractive Index: 1.5100
Odor: Characteristic
Boiling Point: 201°
Beilstein Reference: 2242511
MDL No.: MFCD00004631
Formula:
C₇H₇FO Boiling Pt: 201 °C Density: 1.166 Flash Pt: 90 °C(194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004631 CAS Number: 456-47-3 EINECS: 207-265-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...