3-Amino-5-nitrobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A12508-06 A12508-14
CAAAA12508-03EA 113.41 CAD
CAAAA12508-03 CAAAA12508-06 CAAAA12508-14
3-Amino-5-nitrobenzoic acid 98%
3-Amino-5-nitrobenzoic acid

Purity: 98% CAS No.: 618-84-8 Molecular Formula: C7H6N2O4 Formula Weight: 182.14 Melting Point: 214-218° Beilstein Reference: 1457935

Formula: C₇H₆N₂O₄
MW: 182.14
Melting Pt: 214…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00017025
CAS Number: 618-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR