3'-Aminoacetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12503-18
A12503-30
CAAAA12503-18EA
49.61
CAD
CAAAA12503-18
CAAAA12503-30
3'-Aminoacetophenone 97%
3'-Aminoacetophenone
Formula:
C₈H₉NO Boiling Pt: 289…291 °C Melting Pt: 94…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007796 CAS Number: 99-03-6 EINECS: 202-722-3 Merck Index: 14,413 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...