Tetrazolium blue chloride
Supplier: Thermo Scientific Chemicals
A12502-03
A12502-06
A12502-14
CAAAA12502-03EA
66.99
CAD
CAAAA12502-03
CAAAA12502-06
CAAAA12502-14
Tetrazolium blue chloride
Tetrazolium blue chloride
CAS No.: 1871-22-3
Molecular Formula: C40H32Cl2N8O2
Formula Weight: 727.66
Melting Point: ca 255° dec.
Beilstein Reference: 3894077
Merck Reference: 14,9244
MDL No.: MFCD00040933
Formula:
C₄₀H₃₂Cl₂N₈O₂ Melting Pt: ∼ 255 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00040933 CAS Number: 1871-22-3 EINECS: 217-488-8 Merck Index: 14,9244 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...