α,α'-Dichloro-o-xylene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12500-18
A12500-30
CAAAA12500-18EA
114.39
CAD
CAAAA12500-18
CAAAA12500-30
α,α'-Dichloro-o-xylene 98%
α,α'-Dichloro-o-xylene
Purity: 98%
CAS No.: 612-12-4
Molecular Formula: C8H8Cl2
Formula Weight: 175.06
Flash Point: 107°(224°F)
Odor: Pungent
Boiling Point: 246°
Beilstein Reference: 2043675
UN No.: 2928
MDL No.: MFCD00000903
Formula:
C₈H₈Cl₂ Boiling Pt: 246 °C Melting Pt: 53…57 °C Density: 1.393 g/cm³ (0 °C) Flash Pt: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000903 CAS Number: 612-12-4 EINECS: 210-291-8 UN: 2928 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...