5-Chloro-2-nitrobenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A12493-06 A12493-14 A12493-22
CAAAA12493-06EA 29.39 CAD
CAAAA12493-06 CAAAA12493-14 CAAAA12493-22
5-Chloro-2-nitrobenzoic acid 99%
5-Chloro-2-nitrobenzoic acid

Purity: 99% CAS No.: 2516-95-2 Molecular Formula: C7H4ClNO4 Formula Weight: 201.57 Odor: Odorless Melting Point: 136-140° Beilstein Reference: 1963952 MDL No.: MFCD00007290

Formula: C₇H₄ClNO₄
Melting Pt: 136…140 °C
Density: 1,593 g/cm³ (18 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007290
CAS Number: 2516-95-2
EINECS: 219-738-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR