Methyl laurate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dodecylic acid methyl ester, Methyl dodecanoate, Lauric acid methyl ester, Dodecanoic acid methyl ester
A12492-06
A12492-14
A12492-22
CAAAA12492-06EA
68.89
CAD
CAAAA12492-06
CAAAA12492-14
CAAAA12492-22
Methyl laurate 99%
Methyl laurate
Purity: 99%
CAS No.: 111-82-0
Molecular Formula: CH3(CH2)10CO2CH3
Formula Weight: 214.35
Flash Point: 125°(257°F)
Refractive Index: 1.4325
Melting Point: 4-5°
Boiling Point: 261-262°
Beilstein Reference: 1767780
MDL No.: MFCD00008966
Formula:
C₁₃H₂₆O₂ MW: 214.35 g/mol Boiling Pt: 266 Melting Pt: 4.5 Density: 0.870 Flash Pt: 125 °C (257 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008966 CAS Number: 111-82-0 EINECS: 203-911-3 UN: 3082 ADR: 9 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...