O-Acetylsalicylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Acetoxybenzoic acid, Acetylsalicylic acid, Aspirin, Aspirin®

A12488-22 A12488-0E A12488-36 A12488-36
CAAAAA12488-22EA 46.93 CAD
CAAAAA12488-22 CAAAA12488-0E CAAAA12488-36 AAA12488-36
O-Acetylsalicylic acid 99%
o-Acetylsalicylic acid

Purity: 99% CAS No.: 50-78-2 Molecular Formula: C9H8O4 Formula Weight: 180.15 Flash Point: 250°(482°F) Melting Point: 133-140° Beilstein Reference: 779271 Merck Reference: 14,851 MDL No.: MFCD00002430

Formula: 2-(CH₃CO₂)C₆H₄CO₂H
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 133…140 °C
Density: 1.400
Flash Pt: 250 °C(482 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002430
CAS Number: 50-78-2
EINECS: 200-064-1
Merck Index: 14,851
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fcb60a4ee505b596cef38216a85e034985091e0&vid=5f8d5cfeb3039783869a9320ec840b8ec12c33ea&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR