Tris(4-chlorophenyl)phosphine 97%

Supplier: Thermo Scientific Chemicals
Warning

A12487-14 A12487-03 A12487-06
CAAAAA12487-14EA 1531.02 CAD
CAAAAA12487-14 CAAAA12487-03 CAAAA12487-06
Tris(4-chlorophenyl)phosphine 97%
Tris(4-chlorophenyl)phosphine

Purity: 97% CAS No.: 1159-54-2 Molecular Formula: C18H12Cl3P Formula Weight: 365.63 Melting Point: 102-103° Beilstein Reference: 751458

Formula: C₁₈H₁₂Cl₃P
Boiling Pt: 431 °C (1013 hPa)
Melting Pt: 102…103 °C
Density: 1.449 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013639
CAS Number: 1159-54-2
EINECS: 214-596-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR