Acetyl bromide 98+%

Supplier: Thermo Scientific Chemicals
Danger

A12485-14 A12485-22 A12485-36
CAAAAA12485-14EA 59.41 CAD
CAAAAA12485-14 CAAAA12485-22 CAAAA12485-36
Acetyl bromide 98+%
Acetyl bromide

Purity: 98+% CAS No.: 506-96-7 Molecular Formula: C2H3BrO Formula Weight: 122.95 Flash Point: 75°(167°F) Physical Form: Liquid Density: 1.660 Refractive Index: 1.4500 Sensitivity: Moisture Sensitive Appearance: Colorless Odor: Acrid Melting Point: -96° Boiling Point: 75-77° Beilstein Reference: 1697546 Merck Reference: 14,82 UN No.: 1716 MDL No.: MFCD00000114

Formula: C₂H₃BrO
MW: 122.95 g/mol
Boiling Pt: 75…77 °C
Melting Pt: –96 °C
Density: 1.660
Flash Pt: >75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000114
CAS Number: 506-96-7
EINECS: 208-061-7
UN: 1716
ADR: 8 II
Merck Index: 14,82

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR